Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289395.1 | NP_001276324.1 | 10 | 908 | ||
NM_003322.4 | NP_003313.3 | 11 | 1067 | |||
GenBank mRNA | AB593123.1 | - | 10 | 1064 | ||
AB593124.1 | - | 11 | 1053 | |||
BC032714.1 | - | 10 | 893 | 5495136 | ||
BC065261.1 | - | 10 | 876 | 6144003 | ||
U82468.1 | - | 11 | 1059 |