Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003323.2 | NP_003314.2 | 10 | 1269 | ||
XM_006723347.3 | XP_006723410.1 | 10 | 1269 | |||
XM_006723348.3 | XP_006723411.1 | 10 | 1213 | |||
XM_011527252.2 | XP_011525554.1 | 11 | 1373 | |||
XM_011527253.2 | XP_011525555.1 | 10 | 1260 | |||
XM_011527254.2 | XP_011525556.1 | 10 | 1269 | |||
XM_011527255.2 | XP_011525557.1 | 9 | 1217 | |||
XM_011527256.2 | XP_011525558.1 | 8 | 1184 | |||
GenBank mRNA | BC026070.2 | - | 10 | 1263 | 4837710 | |
U82469.1 | - | 10 | 1211 |