Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 5, 6 | 608 | ||
XM_011512559.2 | XP_011510861.1 | 6, 7 | 729 | |||
GenBank mRNA | AB023217.3 | - | 5, 6 | 608 |