Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 20 | 2203 | ||
XM_011512559.2 | XP_011510861.1 | 21 | 2324 | |||
XM_017005922.1 | XP_016861411.1 | 11 | 1468 | |||
GenBank mRNA | AB023217.3 | - | 20 | 2203 | ||
AK126801.1 | - | 2 | 138 |