Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228178
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
1
904
NM_001242533.1
NP_001229462.1
2
788
NM_001242534.1
NP_001229463.1
2
650
NM_001242535.1
NP_001229464.1
2
599
NM_001242536.1
NP_001229465.1
2
788
NM_001242537.1
NP_001229466.1
2
599
NM_024311.3
NP_077287.1
2
506
XM_011525236.2
XP_011523538.1
1
168
XM_011525237.2
XP_011523539.2
2
611
XM_011525238.2
XP_011523540.2
2
211
XM_011525239.2
XP_011523541.2
2
181
XM_011525240.2
XP_011523542.2
2
207
XM_011525241.2
XP_011523543.2
2
217
XM_011525242.1
XP_011523544.1
1
168
XM_011525244.1
XP_011523546.1
1
168
XM_011525247.2
XP_011523549.2
2
611
XM_017025065.1
XP_016880554.1
2
239
XM_017025066.1
XP_016880555.1
2
181
XM_017025067.1
XP_016880556.1
2
239
XM_017025068.1
XP_016880557.1
2
611
XM_017025069.1
XP_016880558.1
1
398
XM_017025070.1
XP_016880559.1
1
398
GenBank mRNA
AF015183.1
-
1
1018
AF015184.1
-
2
218
AF015185.1
-
2
405
AF015186.1
-
2
362
AF015187.1
-
2
362
AF015188.1
-
3
1189
AF015189.1
-
1
2094
AF015190.1
-
2
650
AK000233.1
-
1
286
AK025849.1
-
1
41
AK308058.1
-
2
243
BC002753.2
-
2
306
3629029
BC062563.1
-
2
399
5140496
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108308, XM_006247811, XM_006247810, XM_006247809.This siRNA also targets the following transcripts in Mouse: NM_178620, BC137971, BC137970, BC100547, BC016209, and others.
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0