Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014997.3 | NP_055812.1 | 4 | 630 | ||
XM_005250233.4 | XP_005250290.1 | 3 | 519 | |||
GenBank mRNA | BC009977.2 | - | 2 | 43 | ||
BC036464.2 | - | 4 | 612 | 4828273 | ||
BC044884.1 | - | 4 | 632 | 5273087 | ||
D87454.1 | - | 4 | 372 |