Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085437.2 | NP_001078906.2 | 5 | 2192 | ||
NM_001282921.1 | NP_001269850.1 | 5 | 1685 | |||
NM_024861.3 | NP_079137.3 | 5 | 1628 | |||
XM_011511877.1 | XP_011510179.1 | 6 | 2184 | |||
GenBank mRNA | AK026324.1 | - | 5 | 1625 | ||
AK096434.1 | - | 5 | 1685 |