Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253845.1 | NP_001240774.1 | 3 | 1382 | ||
XM_017028940.1 | XP_016884429.1 | 2 | 1400 | |||
GenBank mRNA | AK024788.1 | - | 2 | 1605 | ||
AK090635.1 | - | 2 | 1400 | |||
AK225097.1 | - | 2 | 1605 |