Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144876.1 | NP_001138348.1 | 2 | 231 | ||
NM_001308235.1 | NP_001295164.1 | 2 | 341 | |||
NM_001308236.1 | NP_001295165.1 | 2 | 164 | |||
NM_024872.2 | NP_079148.2 | 2 | 120 | |||
XM_005265985.3 | XP_005266042.1 | 2 | 177 | |||
XM_005265987.1 | XP_005266044.1 | 2 | 177 | |||
XM_011534657.2 | XP_011532959.1 | 2 | 187 | |||
XM_017009883.1 | XP_016865372.1 | 1 | 158 | |||
XM_017009884.1 | XP_016865373.1 | 1 | 158 | |||
GenBank mRNA | AK026223.1 | - | 2 | 115 | ||
AK307300.1 | - | 2 | 187 | |||
AL583057.3 | - | 2 | 295 | |||
BI489608.1 | - | 2 | 141 | |||
CF138017.1 | - | 2 | 272 | |||
DA674730.1 | - | 2 | 121 | |||
DA931137.1 | - | 2 | 231 |