Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303247.1 | NP_001290176.1 | 11 | 1164 | ||
NM_001303248.1 | NP_001290177.1 | 9 | 779 | |||
NM_025234.2 | NP_079510.1 | 11 | 1058 | |||
XM_011522094.1 | XP_011520396.1 | 11 | 1038 | |||
GenBank mRNA | AF100786.1 | - | 11 | 919 | ||
AF127799.1 | - | 10 | 907 | |||
AF309553.1 | - | 10 | 903 | |||
AK024754.1 | - | 11 | 930 | |||
AK025098.2 | - | 1 | 3369 | |||
BC010080.2 | - | 11 | 922 | 4329599 | ||
BU528743.1 | - | 9 | 642 | |||
CR457333.1 | - | 9 | 854 |