Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014999.3 | NP_055814.1 | 3 | 556 | ||
GenBank mRNA | AF091035.1 | - | 3 | 544 | ||
BC009109.1 | - | 2 | 140 | |||
BC021901.1 | - | 3 | 545 | 4342334 | ||
BC092475.1 | - | 3 | 545 | 30339933 | ||
D42087.1 | - | 2 | 97 |