Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013836.1 | NP_001013858.1 | 4 | 492 | ||
NM_001013837.1 | NP_001013859.1 | 4 | 489 | |||
NM_001304523.1 | NP_001291452.1 | 3 | 313 | |||
NM_003550.2 | NP_003541.2 | 4 | 529 | |||
XM_011515567.1 | XP_011513869.1 | 2 | 236 | |||
XM_011515568.2 | XP_011513870.1 | 2 | 236 | |||
XM_017012690.1 | XP_016868179.1 | 4 | 707 | |||
GenBank mRNA | AF083811.1 | - | 4 | 475 | ||
AF123318.1 | - | 2 | 226 | |||
AK310831.1 | - | 4 | 489 | |||
AK314227.1 | - | 3 | 310 | |||
BC009964.2 | - | 3 | 439 | 4299982 | ||
BM818088.1 | - | 4 | 492 | |||
BQ684437.1 | - | 4 | 475 | |||
U33822.1 | - | 3 | 405 |