Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 10 | 1537 | ||
NM_033624.2 | NP_296373.1 | 10 | 1558 | |||
XM_017019037.1 | XP_016874526.1 | 9 | 1383 | |||
XM_017019038.1 | XP_016874527.1 | 9 | 1362 | |||
XM_017019039.1 | XP_016874528.1 | 9 | 1153 | |||
GenBank mRNA | AB020682.1 | - | 10 | 1499 | ||
AF174601.1 | - | 10 | 1498 | |||
AK001699.1 | - | 10 | 1521 | |||
AK074714.1 | - | 7 | 1006 | |||
AK308141.1 | - | 11 | 1443 | |||
AK314573.1 | - | 10 | 1510 | |||
AL136899.1 | - | 10 | 1371 | |||
BC034045.1 | - | 11 | 1464 | 4815947 | ||
BC042333.1 | - | 10 | 1257 | 5195515 | ||
BC091496.1 | - | 10 | 1496 |