Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 9 | 804 | ||
XM_011537300.2 | XP_011535602.1 | 8 | 463 | |||
XM_017021751.1 | XP_016877240.1 | 5 | 364 |