Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030771.1 | NP_110398.1 | 1, 2 | 421 | ||
NM_080654.2 | NP_542385.1 | 1, 2 | 421 | |||
GenBank mRNA | AF155108.1 | - | 1, 2 | 1047 | ||
AF382034.1 | - | 1, 2 | 310 | |||
AK308494.1 | - | 2, 3 | 369 | |||
AK313358.1 | - | 1, 2 | 388 | |||
AY598334.1 | - | 1, 2 | 367 | |||
BC008496.1 | - | 1, 2 | 389 | 4276656 | ||
BC029421.1 | - | 1, 2 | 96 | |||
BU595712.1 | - | 424 |