Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGPS1 SIRNA Assays ›
Gene Symbol:
RALGPS1
Gene Name:
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases:
RALGEF2, RALGPS1A
Chromosome Location:
Chr.9: 126914774 - 127223166 on Build GRCh38
UniGene ID:
Hs.432842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228522
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGPS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190728.1
NP_001177657.1
8
752
NM_001190729.1
NP_001177658.1
9
991
NM_001190730.1
NP_001177659.1
9
991
NM_001322320.1
NP_001309249.1
9
954
NM_001322321.1
NP_001309250.1
9
991
NM_001322322.1
NP_001309251.1
9
991
NM_001322323.1
NP_001309252.1
8
901
NM_001322324.1
NP_001309253.1
9
991
NM_001322325.1
NP_001309254.1
9
991
NM_014636.2
NP_055451.1
9
954
XM_006717328.3
XP_006717391.2
10
965
XM_011519225.1
XP_011517527.1
9
914
XM_011519226.1
XP_011517528.1
9
914
XM_011519227.1
XP_011517529.1
8
863
XM_011519228.1
XP_011517530.1
9
914
XM_011519229.1
XP_011517531.1
10
971
XM_011519230.2
XP_011517532.1
10
965
XM_011519231.1
XP_011517533.1
9
914
XM_011519232.1
XP_011517534.1
9
914
XM_011519233.1
XP_011517535.1
8
752
XM_011519234.2
XP_011517536.1
9
1048
XM_011519235.2
XP_011517537.1
10
1139
XM_011519236.1
XP_011517538.1
8
752
XM_011519238.2
XP_011517540.1
9
914
XM_017015342.1
XP_016870831.1
9
914
XM_017015343.1
XP_016870832.1
10
958
XM_017015344.1
XP_016870833.1
7
731
XM_017015345.1
XP_016870834.1
9
914
XM_017015346.1
XP_016870835.1
8
752
XM_017015347.1
XP_016870836.1
9
811
XM_017015348.1
XP_016870837.1
9
985
XM_017015349.1
XP_016870838.1
6
599
XM_017015350.1
XP_016870839.1
8
752
GenBank mRNA
AB002349.1
-
9
954
AF221098.1
-
8
752
AK299149.1
-
9
932
BC032372.1
-
9
914
5163332
BC033708.1
-
9
984
3901593
BX648170.1
-
2
849
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
regulation of Ral protein signal transduction
intracellular signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Ral guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0