Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014654.3 | NP_055469.3 | 3 | 525 | ||
XM_011542463.1 | XP_011540765.1 | 3 | 460 | |||
XM_011542464.2 | XP_011540766.1 | 3 | 730 | |||
XM_011542466.1 | XP_011540768.1 | 3 | 367 | |||
GenBank mRNA | AB007937.1 | - | 1 | 1807 | ||
AF248634.1 | - | 3 | 532 | |||
AK295698.1 | - | 2 | 270 | |||
KU178484.1 | - | 2 | 262 | |||
KU178485.1 | - | 1 | 304 |