Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128219.2 | NP_001121691.1 | 5 | 2225 | ||
NM_001128220.2 | NP_001121692.1 | 4 | 1600 | |||
NM_001128221.2 | NP_001121693.1 | 4 | 1814 | |||
NM_001284390.1 | NP_001271319.1 | 7 | 2269 | |||
NM_001284391.1 | NP_001271320.1 | 5 | 1942 | |||
NM_014667.3 | NP_055482.2 | 6 | 2136 | |||
XM_011534267.2 | XP_011532569.1 | 7 | 1911 | |||
XM_011534268.1 | XP_011532570.1 | 5 | 2239 | |||
XM_011534269.1 | XP_011532571.1 | 5 | 1933 | |||
XM_017007541.1 | XP_016863030.1 | 7 | 1934 | |||
XM_017007542.1 | XP_016863031.1 | 6 | 2123 | |||
GenBank mRNA | AK126479.1 | - | 2 | 2858 | ||
AK130542.1 | - | 6 | 1932 | |||
D50911.3 | - | 6 | 1884 |