Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161576.2 | NP_001155048.2 | 2 | 991 | ||
NM_015589.5 | NP_056404.4 | 2 | 991 | |||
GenBank mRNA | AB028976.1 | - | 1 | 322 | ||
AF429970.1 | - | 2 | 794 | |||
AK024652.1 | - | 2 | 684 | |||
AK025923.1 | - | 1 | 146 | |||
BC057838.1 | - | 2 | 972 | |||
BC121173.1 | - | 2 | 688 | 40122465 | ||
BC121174.1 | - | 2 | 688 | 40122467 | ||
BX648198.1 | - | 6 | 3410 |