Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FNBP1 SIRNA Assays ›
Gene Symbol:
FNBP1
Gene Name:
formin binding protein 1
Gene Aliases:
FBP17
Chromosome Location:
Chr.9: 129887187 - 130053846 on Build GRCh38
UniGene ID:
Hs.189409
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22915
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FNBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015033.2
NP_055848.1
15
1838
XM_005251815.2
XP_005251872.1
16
1868
XM_005251820.2
XP_005251877.1
15
1814
XM_005251821.2
XP_005251878.1
15
1811
XM_005251822.2
XP_005251879.1
14
1799
XM_005251823.2
XP_005251880.1
14
1796
XM_005251824.2
XP_005251881.1
14
1796
XM_005251825.2
XP_005251882.1
15
1781
XM_005251826.2
XP_005251883.1
15
1781
XM_005251827.2
XP_005251884.1
14
1769
XM_005251828.2
XP_005251885.1
14
1766
XM_005251829.2
XP_005251886.1
15
1724
XM_005251830.2
XP_005251887.1
14
1712
XM_005251831.2
XP_005251888.1
14
1631
XM_005251832.2
XP_005251889.1
13
1616
XM_005251833.2
XP_005251890.1
13
1613
XM_005251834.2
XP_005251891.1
13
1613
XM_006717016.2
XP_006717079.1
16
1811
XM_011518399.2
XP_011516701.1
16
1898
XM_011518400.2
XP_011516702.1
16
1895
XM_011518401.1
XP_011516703.1
16
1898
XM_011518402.1
XP_011516704.1
16
1895
XM_017014487.1
XP_016869976.1
16
1808
XM_017014488.1
XP_016869977.1
15
1784
XM_017014489.1
XP_016869978.1
15
1727
XM_017014490.1
XP_016869979.1
15
1727
XM_017014491.1
XP_016869980.1
15
1724
XM_017014492.1
XP_016869981.1
16
2184
XM_017014493.1
XP_016869982.1
14
1628
XM_017014494.1
XP_016869983.1
14
1628
GenBank mRNA
AB011126.1
-
15
1792
AF265550.1
-
15
1807
AK000975.1
-
13
1500
AK001616.1
-
13
1462
AK023681.1
-
15
1724
AK293743.1
-
6
601
BC101755.1
-
15
1680
8069261
BC143513.1
-
14
1665
9052022
BC143514.1
-
15
1650
9052023
BC143515.1
-
13
1482
9052024
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
vesicle-mediated transport
positive regulation of GTPase activity
ARF guanyl-nucleotide exchange factor activity
protein binding
lipid binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0