Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015092.4 | NP_055907.3 | 27 | 4272 | ||
XM_005255182.3 | XP_005255239.1 | 27 | 4285 | |||
XM_005255183.3 | XP_005255240.1 | 28 | 4343 | |||
XM_005255184.3 | XP_005255241.1 | 26 | 4129 | |||
XM_011545769.2 | XP_011544071.1 | 27 | 4285 | |||
XM_011545770.1 | XP_011544072.1 | 15 | 2082 | |||
XM_017023066.1 | XP_016878555.1 | 27 | 4095 | |||
XM_017023067.1 | XP_016878556.1 | 15 | 2145 | |||
GenBank mRNA | AB061371.1 | - | 28 | 4224 | ||
AF186377.1 | - | 2 | 253 | |||
AF395444.1 | - | 25 | 3579 | |||
AY014957.1 | - | 15 | 2151 |
Gene Symbol: SMG1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027154.1 | 27 | 4292 | |||
GenBank mRNA | AK300040.1 | - | 13 | 1767 | ||
BC090929.1 | - | 15 | 2201 |
Gene Symbol: SMG1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027155.2 | 27 | 4333 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627362.1 | - | 2 | 926 |
Gene Symbol: SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135312.1 | 17 | 2419 |
Gene Symbol: SMG1P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033959.1 | 3 | 1392 | |||
GenBank mRNA | AK128245.1 | - | 3 | 1392 |