Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015036.2 | NP_055851.1 | 2 | 528 | ||
GenBank mRNA | AB020637.1 | - | 2 | 479 | ||
AK026560.1 | - | 1 | 41 | |||
AK291673.1 | - | 2 | 528 | |||
BC026191.1 | - | 2 | 495 | 4816880 | ||
BC071171.1 | - | 2 | 479 | 5755016 |