Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137664.1 | NP_001131136.1 | 8 | 1110 | ||
NM_016238.2 | NP_057322.2 | 8 | 1110 | |||
XM_005253903.3 | XP_005253960.1 | 7 | 976 | |||
XM_011538454.2 | XP_011536756.1 | 8 | 1338 | |||
XM_017019425.1 | XP_016874914.1 | 8 | 1110 | |||
XM_017019426.1 | XP_016874915.1 | 8 | 1105 | |||
XM_017019427.1 | XP_016874916.1 | 8 | 1904 | |||
XM_017019428.1 | XP_016874917.1 | 7 | 993 | |||
GenBank mRNA | AF191340.1 | - | 8 | 1007 | ||
AK021809.1 | - | 8 | 1089 | |||
AL137586.1 | - | 2 | 164 | |||
BC002941.2 | - | 3 | 254 | |||
BC009498.2 | - | 6 | 680 | |||
BC017316.2 | - | 4 | 456 | |||
BC040130.1 | - | 8 | 1025 | |||
BC098264.1 | - | 8 | 1006 | |||
BC098295.1 | - | 8 | 1006 | |||
BC098362.1 | - | 8 | 1006 | |||
BC099732.1 | - | 8 | 1006 | |||
BC111798.1 | - | 8 | 1006 | |||
BC141848.1 | - | 8 | 1060 | 8860346 | ||
BC148236.2 | - | 8 | 1007 | 40024981 |