Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323916.1 | NP_001310845.1 | 5 | 721 | ||
NM_020840.2 | NP_065891.1 | 5 | 716 | |||
XM_005263156.1 | XP_005263213.1 | 5 | 716 | |||
XM_005263158.1 | XP_005263215.1 | 5 | 516 | |||
XM_005263160.3 | XP_005263217.1 | 5 | 716 | |||
XM_011532154.2 | XP_011530456.1 | 5 | 1496 | |||
XM_017008487.1 | XP_016863976.1 | 5 | 716 | |||
GenBank mRNA | AB040883.1 | - | 5 | 573 | ||
AL713802.1 | - | 5 | 517 | |||
BC016638.1 | - | 5 | 721 |