Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015044.4 | NP_055859.1 | 7 | 719 | ||
XM_017023074.1 | XP_016878563.1 | 8 | 4408 | |||
XM_017023075.1 | XP_016878564.1 | 7 | 722 | |||
GenBank mRNA | AB029003.1 | - | 4 | 349 | ||
AF165531.1 | - | 7 | 657 | |||
AF190863.1 | - | 7 | 663 | |||
AF233522.1 | - | 7 | 662 | |||
AF323754.1 | - | 8 | 801 | |||
AK024619.1 | - | 7 | 718 | |||
AK308425.1 | - | 7 | 706 | |||
BC000284.1 | - | 7 | 701 | 3356490 |