Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016558.3 | NP_057642.1 | 3 | 1351 | ||
NM_033630.2 | NP_361012.2 | 2 | 882 | |||
GenBank mRNA | AF204271.1 | - | 1 | 847 | ||
AF207829.1 | - | 2 | 762 | |||
BC000785.2 | - | 2 | 746 | 3508136 |