Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015183.2 | NP_055998.1 | 3 | 161 | ||
NM_198828.2 | NP_942123.1 | 5 | 1004 | |||
XM_011543382.2 | XP_011541684.1 | 6 | 1060 | |||
XM_017009447.1 | XP_016864936.1 | 6 | 1060 | |||
GenBank mRNA | AY830839.1 | - | 3 | 162 | ||
BC033215.1 | - | 5 | 972 | 5015614 |