Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001084392.1 | NP_001077861.1 | 2, 3 | 360 | ||
NM_001355.3 | NP_001346.1 | 3, 4 | 411 | |||
GenBank mRNA | AK292044.1 | - | 2, 3 | 304 | ||
BC005971.1 | - | 2, 3 | 308 | 4082777 | ||
BC015508.2 | - | 2, 3 | 329 | 3902943 | ||
BT006970.1 | - | 2, 3 | 283 | |||
CR456431.1 | - | 2, 3 | 306 | |||
U49785.1 | - | 3, 4 | 409 | |||
U84143.1 | - | 2, 3 | 297 |