Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002160.3 | NP_002151.2 | 5, 6 | 2660 | ||
XM_005251972.3 | XP_005252029.1 | 5, 6 | 2610 | |||
XM_005251973.3 | XP_005252030.1 | 5, 6 | 2610 | |||
XM_005251974.3 | XP_005252031.1 | 5, 6 | 2610 | |||
XM_005251975.3 | XP_005252032.1 | 5, 6 | 2610 | |||
XM_006717096.3 | XP_006717159.1 | 5, 6 | 2610 | |||
XM_006717097.3 | XP_006717160.1 | 5, 6 | 2610 | |||
XM_006717098.3 | XP_006717161.1 | 5, 6 | 2610 | |||
XM_006717101.3 | XP_006717164.1 | 5, 6 | 2610 | |||
XM_011518625.2 | XP_011516927.1 | 5, 6 | 2610 | |||
XM_011518626.2 | XP_011516928.1 | 5, 6 | 2610 | |||
XM_011518628.2 | XP_011516930.1 | 5, 6 | 2610 | |||
XM_011518629.2 | XP_011516931.1 | 5, 6 | 2610 | |||
XM_017014678.1 | XP_016870167.1 | 5, 6 | 2610 | |||
XM_017014679.1 | XP_016870168.1 | 5, 6 | 2610 | |||
XM_017014680.1 | XP_016870169.1 | 5, 6 | 2610 | |||
XM_017014681.1 | XP_016870170.1 | 5, 6 | 2610 | |||
GenBank mRNA | AB210038.1 | - | 5, 6 | 2536 | ||
AK304015.1 | - | 5, 6 | 2556 | |||
BC151843.1 | - | 5, 6 | 2528 | 8860362 | ||
GU473235.1 | - | 4, 5 | 2248 | |||
M24630.1 | - | 3, 4 | 958 | |||
M55618.1 | - | 4, 5 | 2383 | |||
X56160.1 | - | 4, 5 | 2299 | |||
X78565.1 | - | 5, 6 | 2561 |