Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139514.1 | NP_001132986.1 | 5, 6 | 1227 | ||
NM_001139515.1 | NP_001132987.1 | 6, 7 | 652 | |||
NM_053281.3 | NP_444511.1 | 6, 7 | 1266 | |||
XM_011530846.2 | XP_011529148.1 | 6, 7 | 1265 | |||
XM_011530847.2 | XP_011529149.1 | 6, 7 | 1265 | |||
XM_011530848.2 | XP_011529150.1 | 6, 7 | 1265 | |||
XM_017029254.1 | XP_016884743.1 | 5, 6 | 1226 | |||
XM_017029255.1 | XP_016884744.1 | 5, 6 | 1226 | |||
XM_017029256.1 | XP_016884745.1 | 5, 6 | 1226 | |||
GenBank mRNA | AF428101.1 | - | 6, 7 | 1096 | ||
AK055953.1 | - | 5, 6 | 613 | |||
AK091842.1 | - | 5, 6 | 1220 | |||
AK290596.1 | - | 6, 7 | 1266 | |||
AK294498.1 | - | 1, 2 | 61 | |||
AK295648.1 | - | 1, 2 | 62 | |||
AK298787.1 | - | 6, 7 | 652 | |||
BC114950.1 | - | 6, 7 | 1175 | 40006598 |