Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105521.3 | NP_001098991.1 | 20, 21 | 2660 | ||
NM_001323086.1 | NP_001310015.1 | 20, 21 | 2666 | |||
NM_001323087.1 | NP_001310016.1 | 20, 21 | 2666 | |||
NM_001323088.1 | NP_001310017.1 | 20, 21 | 2660 | |||
NM_001323089.1 | NP_001310018.1 | 19, 20 | 2438 | |||
NM_001323090.1 | NP_001310019.1 | 20, 21 | 2660 | |||
XM_005252674.4 | XP_005252731.1 | 20, 21 | 4654 | |||
XM_005252676.4 | XP_005252733.1 | 19, 20 | 4438 | |||
XM_011539676.2 | XP_011537978.1 | 20, 21 | 2912 | |||
XM_011539682.2 | XP_011537984.1 | 19, 20 | 4444 | |||
XM_017016095.1 | XP_016871584.1 | 20, 21 | 4660 | |||
XM_017016096.1 | XP_016871585.1 | 20, 21 | 2905 | |||
XM_017016097.1 | XP_016871586.1 | 19, 20 | 4432 | |||
GenBank mRNA | AK126964.1 | - | 10, 11 | 947 | ||
AL832756.1 | - | 10, 11 | 1808 |