Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302819.1 | NP_001289748.1 | 1, 2 | 542 | ||
NM_015372.2 | NP_056187.1 | 2, 3 | 333 | |||
GenBank mRNA | AJ704765.1 | - | 2, 3 | 284 | ||
AK294232.1 | - | 1, 2 | 512 | |||
AK315264.1 | - | 2, 3 | 331 | |||
AL050256.1 | - | 2, 3 | 321 |