Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015050.2 | NP_055865.1 | 4 | 621 | ||
XM_005248955.3 | XP_005249012.1 | 4 | 551 | |||
GenBank mRNA | AK000348.1 | - | 599 | |||
AK292564.1 | - | 4 | 475 | |||
BC031890.1 | - | 4 | 515 | 4944457 | ||
D43949.2 | - | 4 | 511 |