Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100399.1 | NP_001093869.1 | 16 | 2262 | ||
NM_001100400.1 | NP_001093870.1 | 16 | 2262 | |||
XM_011513672.2 | XP_011511974.1 | 16 | 1875 | |||
XM_017007928.1 | XP_016863417.1 | 16 | 1837 | |||
GenBank mRNA | AB014548.1 | - | 4 | 266 | ||
AB621803.1 | - | 16 | 2276 | |||
AF294791.1 | - | 16 | 1833 | |||
AK098331.1 | - | 16 | 1846 | |||
AY550968.1 | - | 15 | 1759 | |||
BC041361.1 | - | 16 | 2266 | |||
BC114218.1 | - | 16 | 1958 | 5579832 | ||
BC126225.1 | - | 15 | 1672 | 8991941 | ||
CR749425.1 | - | 15 | 1631 |