Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128591.1 | NP_001122063.1 | 3 | 414 | ||
NM_001128592.1 | NP_001122064.1 | 4 | 531 | |||
XM_011514591.2 | XP_011512893.1 | 4 | 966 | |||
GenBank mRNA | AK126623.1 | - | 2 | 4205 | ||
AL389983.1 | - | 1 | 1375 | |||
BM738331.1 | - | 4 | 508 | |||
BX648600.1 | - | 1 | 4145 | |||
CN429483.1 | - | 3 | 382 |