Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM153A SIRNA Assays ›
Gene Symbol:
FAM153A
Gene Name:
family with sequence similarity 153 member A
Gene Aliases:
NY-REN-7
Chromosome Location:
Chr.5: 177694247 - 177783518 on Build GRCh38
UniGene ID:
Hs.368516
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229732
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM153A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173663.3
NP_775934.3
12
587
XM_005265889.3
XP_005265946.1
13
775
XM_006714848.3
XP_006714911.1
13
747
XM_006714849.3
XP_006714912.1
12
572
XM_011534520.2
XP_011532822.1
13
775
XM_011534521.2
XP_011532823.1
14
812
XM_011534522.2
XP_011532824.1
14
896
XM_011534523.2
XP_011532825.1
13
837
XM_017009352.1
XP_016864841.1
13
775
XM_017009353.1
XP_016864842.1
13
775
XM_017009354.1
XP_016864843.1
13
775
XM_017009355.1
XP_016864844.1
13
775
XM_017009356.1
XP_016864845.1
13
775
XM_017009357.1
XP_016864846.1
13
775
XM_017009358.1
XP_016864847.1
13
775
XM_017009359.1
XP_016864848.1
13
775
XM_017009360.1
XP_016864849.1
13
775
XM_017009361.1
XP_016864850.1
13
775
XM_017009362.1
XP_016864851.1
12
619
XM_017009363.1
XP_016864852.1
14
696
XM_017009364.1
XP_016864853.1
14
853
XM_017009365.1
XP_016864854.1
14
696
XM_017009366.1
XP_016864855.1
14
865
XM_017009367.1
XP_016864856.1
13
743
XM_017009368.1
XP_016864857.1
13
821
XM_017009369.1
XP_016864858.1
13
760
XM_017009370.1
XP_016864859.1
13
760
XM_017009371.1
XP_016864860.1
13
760
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0