Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 15 | 2000 | ||
NM_001265611.1 | NP_001252540.1 | 14 | 1982 | |||
NM_014994.2 | NP_055809.2 | 14 | 1982 | |||
NR_049761.1 | 13 | 1792 | ||||
NR_049762.1 | 12 | 1743 | ||||
XM_006720438.1 | XP_006720501.1 | 13 | 1799 | |||
XM_006720439.2 | XP_006720502.1 | 1 | 78 | |||
XM_011521383.1 | XP_011519685.1 | 14 | 1817 | |||
XM_011521384.2 | XP_011519686.1 | 15 | 1964 | |||
XM_017022017.1 | XP_016877506.1 | 13 | 2093 | |||
GenBank mRNA | BC113983.1 | - | 14 | 1874 | 40083186 |