Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297714.1 | NP_001284643.1 | 10 | 1516 | ||
NM_015055.3 | NP_055870.2 | 11 | 1690 | |||
XM_005252830.4 | XP_005252887.1 | 9 | 1591 | |||
GenBank mRNA | AB014540.1 | - | 11 | 1642 | ||
AF134894.1 | - | 11 | 1661 | |||
AF161439.1 | - | 5 | 529 | |||
AF210818.1 | - | 11 | 1587 | |||
AK027131.1 | - | 8 | 1117 | |||
AK096859.1 | - | 10 | 1516 | |||
AK304104.1 | - | 11 | 1554 | |||
BC000616.2 | - | 11 | 1663 | 3346240 |