Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039775.3 | NP_001034864.2 | 10 | 3888 | ||
XM_005245918.2 | XP_005245975.1 | 10 | 3874 | |||
XM_011541672.1 | XP_011539974.1 | 9 | 3838 | |||
XM_011541673.2 | XP_011539975.1 | 10 | 4113 | |||
GenBank mRNA | AK095339.1 | - | 9 | 932 | ||
AL137264.1 | - | 4 | 356 | |||
BC136870.1 | - | 6 | 630 | 9020861 |