Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207510.3 | NP_997393.3 | 1 | 638 | ||
GenBank mRNA | AK127160.1 | - | 1 | 638 | ||
AK127236.1 | - | 1 | 1727 | |||
BC042845.1 | - | 1 | 307 | |||
BC137559.1 | - | 1 | 1216 | 9021559 | ||
BC137567.1 | - | 1 | 1216 | 9021567 |