Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 28 | 5555 | ||
XM_005246237.2 | XP_005246294.1 | 30 | 5283 | |||
XM_005246239.2 | XP_005246296.1 | 30 | 5098 | |||
XM_005246240.2 | XP_005246297.1 | 23 | 3300 | |||
XM_005246241.1 | XP_005246298.1 | 23 | 3194 | |||
XM_005246242.4 | XP_005246299.1 | 23 | 3319 | |||
XM_006712189.3 | XP_006712252.1 | 29 | 5288 | |||
XM_006712193.3 | XP_006712256.1 | 23 | 6419 | |||
XM_011510479.2 | XP_011508781.1 | 29 | 5721 | |||
XM_011510483.2 | XP_011508785.2 | 29 | 5318 | |||
XM_017003157.1 | XP_016858646.1 | 29 | 5318 | |||
XM_017003158.1 | XP_016858647.1 | 25 | 4894 | |||
XM_017003159.1 | XP_016858648.1 | 26 | 5077 | |||
XM_017003160.1 | XP_016858649.1 | 13 | 1490 | |||
GenBank mRNA | AB037718.1 | - | 19 | 2612 | ||
AY603755.1 | - | 27 | 5168 |