Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP25-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128598.1 | NP_001122070.1 | 1 | 167 | ||
GenBank mRNA | AB180043.1 | - | 1 | 167 |