Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075100.1 | 7 | 2719 | |||
GenBank mRNA | AB095940.1 | - | 7 | 2719 |
Gene Symbol: NUTM2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI832214.1 | - | 1 | 28 |