Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 12 | 2468 | ||
NM_001322046.1 | NP_001308975.1 | 13 | 3270 | |||
NM_001322048.1 | NP_001308977.1 | 12 | 2525 | |||
NM_001322049.1 | NP_001308978.1 | 11 | 2399 | |||
NM_024622.5 | NP_078898.3 | 13 | 2597 | |||
NR_104020.2 | 12 | 2464 | ||||
XM_006712750.3 | XP_006712813.1 | 12 | 2523 | |||
XM_006712751.3 | XP_006712814.1 | 9 | 2608 | |||
XM_017004911.1 | XP_016860400.1 | 13 | 3910 | |||
XM_017004912.1 | XP_016860401.1 | 8 | 2479 | |||
GenBank mRNA | AB058703.1 | - | 11 | 1779 | ||
AK025554.1 | - | 10 | 1709 | |||
AK055892.1 | - | 12 | 2379 | |||
AK074302.1 | - | 13 | 2488 | |||
AK130797.1 | - | 12 | 2388 | |||
AK225293.1 | - | 10 | 1709 | |||
AL832058.1 | - | 13 | 2480 | |||
BC032687.2 | - | 12 | 1926 | 5581443 |