Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230447
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
21
1999
NM_001178107.1
NP_001171578.1
20
1912
NM_001178108.1
NP_001171579.1
20
1906
NM_001178109.1
NP_001171580.1
19
1816
NM_001178110.1
NP_001171581.1
18
1726
NM_001178113.1
NP_001171584.1
12
1456
NM_001178115.1
NP_001171586.1
6
807
NM_007150.3
NP_009081.2
20
1903
XM_005274730.2
XP_005274787.1
22
2106
XM_005274731.2
XP_005274788.1
22
2103
XM_005274735.2
XP_005274792.1
21
2016
XM_005274737.2
XP_005274794.1
21
2010
XM_005274738.2
XP_005274795.1
21
2007
XM_005274740.2
XP_005274797.1
20
1920
XM_005274741.2
XP_005274798.1
20
1920
XM_005274742.2
XP_005274799.1
20
1917
XM_005274744.2
XP_005274801.1
19
1833
XM_005274745.2
XP_005274802.1
19
1830
XM_005274746.3
XP_005274803.1
18
1749
XM_011531194.1
XP_011529496.1
20
1911
XM_011531195.1
XP_011529497.1
21
1908
XM_017029821.1
XP_016885310.1
23
2238
XM_017029822.1
XP_016885311.1
24
2258
XM_017029823.1
XP_016885312.1
23
2235
XM_017029824.1
XP_016885313.1
22
2154
XM_017029825.1
XP_016885314.1
22
2151
XM_017029826.1
XP_016885315.1
22
2148
XM_017029827.1
XP_016885316.1
22
2145
XM_017029828.1
XP_016885317.1
22
2142
XM_017029829.1
XP_016885318.1
21
2061
XM_017029830.1
XP_016885319.1
21
2055
XM_017029831.1
XP_016885320.1
21
2052
XM_017029832.1
XP_016885321.1
21
2043
XM_017029833.1
XP_016885322.1
20
1977
XM_017029834.1
XP_016885323.1
20
1965
XM_017029835.1
XP_016885324.1
20
1926
GenBank mRNA
AK056517.1
-
9
962
AK095258.1
-
6
779
AK296967.1
-
20
1906
AK301530.1
-
12
1456
AK304323.1
-
16
1469
BX537525.1
-
6
2481
BX640940.1
-
12
1203
DQ508023.1
-
18
1678
DQ508024.1
-
19
1768
DQ508025.1
-
21
1951
DQ508026.1
-
20
1864
Y09538.1
-
12
1184
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0