Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020862.1 | NP_065913.1 | 2 | 3051 | ||
XM_005259102.4 | XP_005259159.1 | 5 | 3251 | |||
XM_017027033.1 | XP_016882522.1 | 5 | 3366 | |||
GenBank mRNA | AB040917.1 | - | 2 | 2840 |