Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166017.1 | NP_001159489.1 | 4 | 720 | ||
NM_145061.5 | NP_659498.4 | 4 | 720 | |||
XM_005266288.3 | XP_005266345.1 | 3 | 506 | |||
XM_011534994.1 | XP_011533296.1 | 4 | 696 | |||
GenBank mRNA | AF361358.1 | - | 4 | 662 | ||
AK090584.1 | - | 4 | 665 | |||
AK307976.1 | - | 5 | 595 | |||
AK314864.1 | - | 4 | 666 | |||
BC013418.2 | - | 3 | 604 | 3604003 | ||
BC048988.1 | - | 4 | 691 | 5267965 |