Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
Gene Name:
ataxin 3
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
UniGene ID:
Hs.532632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230538
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
7
664
NM_001127697.2
NP_001121169.2
6
556
NM_001164779.1
NP_001158251.1
4
346
NM_001164780.1
NP_001158252.1
5
413
NM_001164781.1
NP_001158253.1
6
499
NM_004993.5
NP_004984.2
8
709
NM_030660.4
NP_109376.1
7
544
NR_028453.1
7
623
NR_028454.1
6
458
NR_028455.1
7
623
NR_028456.1
6
458
NR_028457.1
8
714
NR_028458.1
7
600
NR_028459.1
8
709
NR_028461.1
7
621
NR_028462.1
7
600
NR_028463.1
3
258
NR_028464.1
6
556
NR_028465.1
6
578
NR_028466.1
3
258
NR_028467.1
7
624
NR_028469.1
6
458
NR_031765.1
2
125
GenBank mRNA
AB050194.1
-
7
533
AK225884.1
-
8
648
AK314919.1
-
8
661
BC022245.1
-
7
592
BC033711.1
-
8
652
4393766
BC095402.1
-
8
669
30347215
BU190081.1
-
7
643
BU663113.1
-
5
463
GQ176433.1
-
8
662
GQ176434.1
-
8
661
GQ176435.1
-
8
662
GQ176436.1
-
6
509
GQ176437.1
-
6
509
GQ176438.1
-
7
576
GQ176453.1
-
7
552
GQ176454.1
-
8
662
GQ176455.1
-
7
497
GQ176456.1
-
6
411
GQ176457.1
-
6
509
GQ176458.1
-
7
576
GQ176459.1
-
4
299
GQ176462.1
-
8
662
GQ176463.1
-
7
576
GQ176464.1
-
7
576
GQ176465.1
-
3
211
GQ176466.1
-
6
411
GQ176469.1
-
6
509
GQ176470.1
-
6
531
GQ176474.1
-
6
509
GQ176475.1
-
6
411
GQ176476.1
-
8
662
GQ176477.1
-
7
576
GQ176479.1
-
7
576
GQ176481.1
-
7
576
GQ176483.1
-
7
576
GQ176484.1
-
7
576
GQ176485.1
-
6
508
GQ176486.1
-
7
617
GQ176487.1
-
6
509
GQ176488.1
-
8
662
GQ176489.1
-
7
617
GQ176490.1
-
6
543
GQ176492.1
-
7
576
GQ176493.1
-
6
508
GQ176496.1
-
6
411
GQ176497.1
-
7
576
GQ176498.1
-
7
497
GQ176499.1
-
8
662
GQ176503.1
-
6
410
GQ176504.1
-
8
662
GQ176505.1
-
6
411
GQ176506.1
-
6
411
GQ176517.1
-
3
211
GQ176518.1
-
3
211
GQ176519.1
-
7
575
GQ176525.1
-
6
411
GQ176529.1
-
7
573
GQ176531.1
-
8
662
GQ176532.1
-
7
662
GQ176533.1
-
8
662
GQ176534.1
-
7
573
GQ176538.1
-
8
662
GQ176540.1
-
7
550
GQ176542.1
-
4
298
GQ176543.1
-
6
411
GQ176544.1
-
6
411
GQ176545.1
-
4
299
GQ176546.1
-
7
576
GQ176547.1
-
6
411
GQ176548.1
-
8
662
GQ176550.1
-
8
662
GQ176551.1
-
8
660
GQ176552.1
-
8
661
GQ176554.1
-
6
411
GQ176556.1
-
6
411
GQ176559.1
-
6
411
GQ176560.1
-
8
662
GQ176563.1
-
8
662
GQ176565.1
-
8
662
GQ176566.1
-
7
497
GQ176567.1
-
8
662
GQ176568.1
-
8
662
GQ176569.1
-
7
573
GQ176570.1
-
8
662
GQ176571.1
-
6
411
GQ176572.1
-
7
576
GQ176573.1
-
7
497
GQ176576.1
-
7
574
GQ176579.1
-
7
553
GQ176580.1
-
8
662
GQ176608.1
-
6
509
GQ176609.1
-
662
GQ176610.1
-
7
576
GQ176611.1
-
7
576
GQ176613.1
-
3
188
GQ176614.1
-
5
366
GQ176615.1
-
662
GQ176616.1
-
8
662
GQ176617.1
-
7
576
GQ176618.1
-
488
GQ176619.1
-
8
662
GQ176620.1
-
7
576
GQ176621.1
-
8
662
GQ176622.1
-
8
662
GQ176623.1
-
6
411
GQ176624.1
-
7
595
GQ176625.1
-
299
GQ176627.1
-
4
299
GQ176630.1
-
7
576
GQ176631.1
-
6
509
GQ176632.1
-
8
662
GQ176633.1
-
4
299
GQ176636.1
-
5
483
GQ176654.1
-
7
576
GQ176664.1
-
7
576
GQ176665.1
-
7
497
GQ176666.1
-
5
366
GQ176670.1
-
5
464
GQ176673.1
-
8
662
GQ176674.1
-
8
662
GQ176675.1
-
7
497
GQ176676.1
-
6
411
GQ176677.1
-
8
662
GQ176678.1
-
7
576
GQ176679.1
-
7
497
GQ176680.1
-
8
662
GQ176681.1
-
5
421
GQ176682.1
-
7
576
GQ176684.1
-
8
662
GQ176685.1
-
8
661
GQ176687.1
-
8
662
GQ176688.1
-
6
508
GQ176689.1
-
5
366
GQ176690.1
-
7
576
GQ176691.1
-
6
411
GQ176692.1
-
8
662
GQ176695.1
-
6
509
GQ176696.1
-
7
576
GQ176697.1
-
8
662
GQ176700.1
-
7
497
GQ176701.1
-
8
662
GQ176702.1
-
5
366
GQ176703.1
-
8
662
GQ176704.1
-
8
662
GQ176705.1
-
4
299
GQ176706.1
-
7
576
GQ176710.1
-
4
299
GQ176711.1
-
5
366
GQ176712.1
-
7
617
GQ176714.1
-
8
662
GQ176715.1
-
8
584
GQ176716.1
-
6
411
GQ176717.1
-
576
GQ176719.1
-
662
GQ176720.1
-
6
411
GQ176721.1
-
8
662
GQ176722.1
-
7
576
GQ176723.1
-
5
366
GQ176724.1
-
6
509
GQ176725.1
-
8
662
GQ176726.1
-
6
411
GQ176727.1
-
4
299
GQ176728.1
-
8
662
GQ176732.1
-
411
GQ176734.1
-
509
GQ176741.1
-
662
GQ176743.1
-
6
411
GQ176744.1
-
7
617
GQ176745.1
-
662
GQ176746.1
-
8
662
GQ176747.1
-
662
GQ176748.1
-
6
411
GQ176749.1
-
8
662
GQ176750.1
-
7
497
GQ176751.1
-
411
GQ176752.1
-
749
GQ176753.1
-
5
366
GQ176754.1
-
7
497
GQ176755.1
-
7
576
GQ176756.1
-
8
662
GQ176757.1
-
6
509
GQ176758.1
-
6
411
GQ176759.1
-
8
662
GQ176760.1
-
7
576
GQ176761.1
-
5
344
GQ176762.1
-
662
GQ176763.1
-
8
662
GQ176764.1
-
411
GQ176765.1
-
6
411
GQ176766.1
-
8
662
GQ176767.1
-
7
576
GQ176768.1
-
6
411
GQ176769.1
-
5
464
GQ176771.1
-
8
662
GQ176772.1
-
6
509
GQ176773.1
-
6
509
GQ176774.1
-
6
411
GQ176778.1
-
5
366
GQ176779.1
-
6
509
GQ176780.1
-
6
411
GQ176781.1
-
6
509
GQ176782.1
-
6
509
GQ176784.1
-
6
509
GQ176785.1
-
5
344
GQ176786.1
-
6
509
GQ176788.1
-
6
411
GQ176789.1
-
6
509
GQ176790.1
-
6
508
GQ176791.1
-
8
662
GQ176805.1
-
8
638
GQ176806.1
-
8
661
GQ176807.1
-
7
576
GQ176809.1
-
5
464
GQ176810.1
-
7
574
GQ176811.1
-
8
663
GQ176812.1
-
6
509
GQ176813.1
-
7
497
GQ176814.1
-
6
408
GQ176815.1
-
6
510
GQ176819.1
-
7
576
GQ176820.1
-
8
662
GQ176821.1
-
6
410
GQ176822.1
-
7
497
GQ176823.1
-
7
553
GQ176824.1
-
8
662
GQ176826.1
-
8
639
GQ176827.1
-
8
662
GQ176828.1
-
6
411
GQ176829.1
-
8
662
GQ176830.1
-
633
GQ176831.1
-
662
GQ176832.1
-
6
411
GQ176833.1
-
7
576
GQ176834.1
-
575
GQ176835.1
-
7
496
GQ176836.1
-
7
497
GQ176837.1
-
8
662
GQ176839.1
-
411
GQ176842.1
-
576
GQ176844.1
-
6
424
U64820.1
-
8
697
U64821.1
-
8
660
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0