Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135940.1 | NP_001129412.1 | 10 | 1237 | ||
NM_001300911.1 | NP_001287840.1 | 11 | 1368 | |||
NM_006790.2 | NP_006781.1 | 10 | 1719 | |||
XM_017010060.1 | XP_016865549.1 | 11 | 1937 | |||
XM_017010061.1 | XP_016865550.1 | 9 | 1370 | |||
XM_017010062.1 | XP_016865551.1 | 10 | 1806 | |||
GenBank mRNA | AF133820.2 | - | 10 | 1704 | ||
AF144477.1 | - | 10 | 1690 | |||
AK300076.1 | - | 11 | 1348 | |||
AK300088.1 | - | 10 | 1235 | |||
AK312733.1 | - | 10 | 1699 | |||
BC005376.1 | - | 10 | 1699 | 3934230 | ||
KU178476.1 | - | 9 | 1410 | |||
KU178477.1 | - | 3 | 195 |