Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136570.2 | NP_001130042.1 | 5 | 762 | ||
NM_001242936.1 | NP_001229865.1 | 5 | 835 | |||
GenBank mRNA | AK124936.1 | - | 5 | 835 | ||
CR627383.1 | - | 4 | 384 |
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242939.1 | NP_001229868.1 | 5 | 762 | ||
GenBank mRNA | AK090914.1 | - | 3 | 232 | ||
AK092277.1 | - | 5 | 741 | |||
AK296418.1 | - | 4 | 428 |